5-Bromo-2-chlorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

113884-1G 113884-10G
102892-948EA 232.31 USD
102892-948 102892-950
5-Bromo-2-chlorobenzoyl chloride ≥97%
5-Bromo-2-chlorobenzoyl chloride

Hazard: Irritant

Formula: C₇H₃BrCl₂O
MDL Number: MFCD03208978
CAS Number: 21900-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR