6-(Benzyloxy)-5-methylpyrrolo[1,2-f][1,2,4]triazin-4(3H)-one ≥95%

Supplier: Matrix Scientific

115685-500MG 115685-1G 115685-250MG
102897-210EA 1500 USD
102897-210 102897-212 102899-572
6-(Benzyloxy)-5-methylpyrrolo[1,2-f][1,2,4]triazin-4(3H)-one ≥95%
6-(Benzyloxy)-5-methylpyrrolo[1,2-f][1,2,4]triazin-4(3H)-one

Hazard: Irritant

Formula: C₁₄H₁₃N₃O₂
MW: 255.28 g/mol
MDL Number: MFCD11869898
CAS Number: 649736-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR