5-(3-Azetidinyl)-2H-tetrazole hydrochloride ≥95%

Supplier: Matrix Scientific

115684-1G 115684-5G
102899-568EA 1183.08 USD
102899-568 102899-570
5-(3-Azetidinyl)-2H-tetrazole hydrochloride ≥95%
5-(3-Azetidinyl)-2H-tetrazole hydrochloride

Hazard: Irritant

Formula: C₄H₈ClN₅
MDL Number: MFCD21647389
CAS Number: 950691-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR