(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate ≥95%

Supplier: Matrix Scientific

Synonyms: (11bS)-2,6-Bis(3,5-bis(trifluoromethyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

115266-500MG 115266-1G
102895-750EA 1238.46 USD
102895-750 102895-752
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate ≥95%
(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate

Hazard: Irritant

Formula: C₃₆H₁₇F₁₂O₄P
MW: 772.49 g/mol
MDL Number: MFCD08689863
CAS Number: 878111-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR