3-(4-Methylphenyl)-1,4,8-triazaspiro[4.5]dec-3-en-2-one ≥97%

Supplier: Matrix Scientific

115061-500MG 115061-1G
102895-320EA 693.85 USD
102895-320 102895-322
3-(4-Methylphenyl)-1,4,8-triazaspiro[4.5]dec-3-en-2-one ≥97%
3-(4-Methylphenyl)-1,4,8-triazaspiro[4.5]dec-3-en-2-one

Hazard: Irritant

Formula: C₁₄H₁₇N₃O
MW: 243.31 g/mol
MDL Number: MFCD06751103
CAS Number: 887220-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR