[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid ≥97%
Supplier: Matrix Scientific
115056-500MG
115056-1G
102891-276EA
693.85
USD
102891-276
102891-278
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid ≥97%
[7-(3-Methyl-1,2,4-oxadiazol-5-yl)[1,2,4]triazolo[4,3-a]pyridin-3-yl]acetic acid
Hazard: Irritant
Formula:
C₁₁H₉N₅O₃ |
MDL Number:
MFCD20528959 CAS Number: 1338661-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...