2-(4-Chlorophenyl)propionic acid ≥97%

Supplier: Matrix Scientific

114442-1G 114442-10G
102894-046EA 109.23 USD
102894-046 102894-048
2-(4-Chlorophenyl)propionic acid ≥97%
2-(4-Chlorophenyl)propionic acid

Hazard: Irritant

Formula: C₉H₉ClO₂
MDL Number: MFCD00044670
CAS Number: 938-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR