Methyl-2-(benzylamino)acetate ≥97%

Supplier: Matrix Scientific

Synonyms: N-Benzylglycine methyl ester, Methyl 2-(benzylamino)acetate

113955-1G 113955-10G
102893-054EA 192.31 USD
102893-054 102893-056
Methyl-2-(benzylamino)acetate ≥97%
Methyl-2-(benzylamino)acetate

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD03247461
CAS Number: 53386-64-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR