(S)-(+)-2-Hydroxyisovaleric acid ≥97%

Supplier: Matrix Scientific

113854-1G 113854-10G
102892-924EA 60 USD
102892-924 102892-926
(S)-(+)-2-Hydroxyisovaleric acid ≥97%
(S)-(+)-2-Hydroxyisovaleric acid

Hazard: Irritant

Formula: C₅H₁₀O₃
MDL Number: MFCD00066443
CAS Number: 17407-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR