Ethyl tetrahydropyran-4-ylacetate

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(tetrahydro-2H-pyran-4-yl)acetate

113110-1G 113110-5G
102891-502EA 146.15 USD
102891-502 102891-504
Ethyl tetrahydropyran-4-ylacetate
Ethyl tetrahydropyran-4-ylacetate

Hazard: Irritant

Formula: C₉H₁₆O₃
MW: 172.2215 g/mol
MDL Number: MFCD03788560
CAS Number: 103260-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR