5-Amino-1-tosyl-1H-pyrazolo-4-carbonitrile ≥95%

Supplier: Matrix Scientific

115988-1G 115988-25G
102900-164EA 70.77 USD
102900-164 102900-168
5-Amino-1-tosyl-1H-pyrazolo-4-carbonitrile ≥95%
5-Amino-1-tosyl-1H-pyrazolo-4-carbonitrile

Hazard: Irritant

Formula: C₁₁H₁₀N₄O₂S
MDL Number: MFCD04071186
CAS Number: 106368-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR