2,5-Dioxopyrrolidin-1-yltetrahydro-2H-pyran-4-carboxylate ≥95%

Supplier: Matrix Scientific

115986-500MG 115986-5G
102900-152EA 275.38 USD
102900-152 102900-156
2,5-Dioxopyrrolidin-1-yltetrahydro-2H-pyran-4-carboxylate ≥95%
2,5-Dioxopyrrolidin-1-yltetrahydro-2H-pyran-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₃NO₅
MDL Number: MFCD25562901
CAS Number: 1369515-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR