Coumarin-7-pinacolboronate ≥95%

Supplier: Matrix Scientific

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromen-2-one

115979-1G 115979-25G
102900-076EA 880 USD
102900-076 102900-080
Coumarin-7-pinacolboronate ≥95%
Coumarin-7-pinacolboronate

Hazard: Irritant

Formula: C₁₅H₁₇BO₄
MW: 272.11 g/mol
MDL Number: MFCD22989333
CAS Number: 190788-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR