[N-(p-Toluenesulfonyl)imino]phenyliodinane ≥95%

Supplier: Matrix Scientific

115978-1G 115978-5G
102900-034EA 292.31 USD
102900-034 102900-072
[N-(p-Toluenesulfonyl)imino]phenyliodinane ≥95%
[N-(p-Toluenesulfonyl)imino]phenyliodinane

Hazard: Irritant

Formula: C₁₃H₁₄INO₂S
MDL Number: MFCD24850077
CAS Number: 55962-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR