N-(3-Bromo-2-methyl-6-nitrophenyl)-2,2,2-trifluoroacetamide ≥97%

Supplier: Matrix Scientific

115972-1G 115972-10G
102900-064EA 140 USD
102900-064 102900-068
N-(3-Bromo-2-methyl-6-nitrophenyl)-2,2,2-trifluoroacetamide ≥97%
N-(3-Bromo-2-methyl-6-nitrophenyl)-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₉H₆BrF₃N₂O₃
MDL Number: MFCD23703161
CAS Number: 1325729-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR