Methyl-5-(aminomethyl)tetrahydrofuran-2-carboxylate hydrochloride ≥90%

Supplier: Matrix Scientific

115966-500MG 115966-5G
102899-934EA 1318.46 USD
102899-934 102899-938
Methyl-5-(aminomethyl)tetrahydrofuran-2-carboxylate hydrochloride ≥90%
Methyl-5-(aminomethyl)tetrahydrofuran-2-carboxylate hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₃
MDL Number: MFCD23152697
CAS Number: 1304126-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR