11-Azatricyclo[7.3.1.02,7]trideca-2,4,6-triene-9-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

115959-250MG 115959-500MG 115959-1G
102899-642EA 1647.69 USD
102899-642 102899-644 102899-982
11-Azatricyclo[7.3.1.02,7]trideca-2,4,6-triene-9-carboxylic acid ethyl ester ≥95%
11-Azatricyclo[7.3.1.02,7]trideca-2,4,6-triene-9-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₅H₁₉NO₂
MDL Number: MFCD14581516
CAS Number: 1250997-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR