N-[4-(Trifluoromethyl)benzyl]oxetan-3-amine ≥95%

Supplier: Matrix Scientific

115955-500MG 115955-1G 115955-5G
102899-618EA 1238.46 USD
102899-618 102899-620 102899-622
N-[4-(Trifluoromethyl)benzyl]oxetan-3-amine ≥95%
N-[4-(Trifluoromethyl)benzyl]oxetan-3-amine

Hazard: Irritant

Formula: C₁₁H₁₂F₃NO
MDL Number: MFCD18849566
CAS Number: 1344265-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR