(3,4)-trans-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

115953-500MG 115953-5G
102900-248EA 1318.46 USD
102900-248 102900-252
(3,4)-trans-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate ≥95%
(3,4)-trans-tert-Butyl-4-azido-3-fluoropiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₇FN₄O₂
MDL Number: MFCD22628757
CAS Number: 1356634-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR