1-(3-Oxetanyl)-1H-pyrazol-4-amine ≥95%

Supplier: Matrix Scientific

115951-500MG 115951-5G
102900-236EA 1443.08 USD
102900-236 102900-240
1-(3-Oxetanyl)-1H-pyrazol-4-amine ≥95%
1-(3-Oxetanyl)-1H-pyrazol-4-amine

Hazard: Irritant

Formula: C₆H₉N₃O
MDL Number: MFCD21848218
CAS Number: 1338719-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR