[4-(Oxetan-3-yloxy)-3-(trifluoromethyl)phenyl]methanamine ≥95%

Supplier: Matrix Scientific

115946-250MG 115946-1G
102899-954EA 909.23 USD
102899-954 102899-958
[4-(Oxetan-3-yloxy)-3-(trifluoromethyl)phenyl]methanamine ≥95%
[4-(Oxetan-3-yloxy)-3-(trifluoromethyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₁H₁₂F₃NO₂
MDL Number: MFCD20233319
CAS Number: 1349717-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR