4-(Oxetan-3-yloxy)-3-(trifluoromethyl)benzoic acid ≥95%

Supplier: Matrix Scientific

115945-500MG 115945-1G
102898-868EA 1238.46 USD
102898-868 102898-870
4-(Oxetan-3-yloxy)-3-(trifluoromethyl)benzoic acid ≥95%
4-(Oxetan-3-yloxy)-3-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₉F₃O₄
MDL Number: MFCD20233224
CAS Number: 1349716-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR