[4-(Oxetan-3-yloxy)pyridin-2-yl]methanamine ≥95%

Supplier: Matrix Scientific

115943-1G 115943-5G 115943-10G
102898-856EA 2692.31 USD
102898-856 102898-858 102898-860
[4-(Oxetan-3-yloxy)pyridin-2-yl]methanamine ≥95%
[4-(Oxetan-3-yloxy)pyridin-2-yl]methanamine

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD22375705
CAS Number: 1380300-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR