6-(Oxetan-3-yloxy)pyridin-3-amine ≥95%

Supplier: Matrix Scientific

115942-10G 115942-5G
102898-854EA 10080 USD
102898-854 102899-950
6-(Oxetan-3-yloxy)pyridin-3-amine ≥95%
6-(Oxetan-3-yloxy)pyridin-3-amine

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD20233079
CAS Number: 1349716-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR