3,3-Oxetanedimethanamine ≥95%

Supplier: Matrix Scientific

115937-1G 115937-10G
102899-906EA 1978.46 USD
102899-906 102899-910
3,3-Oxetanedimethanamine ≥95%
3,3-Oxetanedimethanamine

Hazard: Irritant

Formula: C₅H₁₂N₂O
MDL Number: MFCD18643361
CAS Number: 23500-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR