2-Oxaspiro[3.5]nonane-7-methanol ≥95%

Supplier: Matrix Scientific

115934-250MG 115934-1G
102899-888EA 1098.46 USD
102899-888 102899-892
2-Oxaspiro[3.5]nonane-7-methanol ≥95%
2-Oxaspiro[3.5]nonane-7-methanol

Hazard: Irritant

Formula: C₉H₁₆O₂
MDL Number: MFCD22628753
CAS Number: 1256546-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR