2-Oxaspiro[3.5]nonane-7-methanamine ≥95%

Supplier: Matrix Scientific

115933-250MG 115933-500MG
102898-850EA 1373.85 USD
102898-850 102898-852
2-Oxaspiro[3.5]nonane-7-methanamine ≥95%
2-Oxaspiro[3.5]nonane-7-methanamine

Hazard: Irritant

Formula: C₉H₁₇NO
MW: 155.24 g/mol
MDL Number: MFCD22628752
CAS Number: 1256667-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR