1-Oxa-6-azaspiro[3.5]nonane ≥95%

Supplier: Matrix Scientific

115929-250MG 115929-500MG 115929-1G
102898-826EA 719.9 USD
102898-826 102898-828 102898-830
1-Oxa-6-azaspiro[3.5]nonane ≥95%
1-Oxa-6-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD13180593
CAS Number: 204388-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR