3-(3-Nitrophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115926-100MG 115926-250MG 115926-500MG
102899-018EA 1593.85 USD
102899-018 102899-020 102899-022
3-(3-Nitrophenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Nitrophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₅
MDL Number: MFCD22546752
CAS Number: 1393552-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR