3-(3-Nitrophenyl)oxetan-3-amine hydrochloride ≥95%
Supplier: Matrix Scientific
115925-250MG
115925-500MG
115925-1G
102899-012EA
2692.31
USD
102899-012
102899-014
102899-016
3-(3-Nitrophenyl)oxetan-3-amine hydrochloride ≥95%
3-(3-Nitrophenyl)oxetan-3-amine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁ClN₂O₃ |
MDL Number:
MFCD22628748 CAS Number: 1393545-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...