5-Nitro-2-(oxetan-3-yloxy)pyrimidine ≥95%

Supplier: Matrix Scientific

115923-1G 115923-5G 115923-10G
102899-006EA 1787.69 USD
102899-006 102899-008 102899-880
5-Nitro-2-(oxetan-3-yloxy)pyrimidine ≥95%
5-Nitro-2-(oxetan-3-yloxy)pyrimidine

Hazard: Irritant

Formula: C₇H₇N₃O₄
MDL Number: MFCD20233117
CAS Number: 1349715-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR