3-(3-Methylphenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115920-100MG 115920-250MG 115920-500MG
102898-808EA 1373.85 USD
102898-808 102898-810 102898-812
3-(3-Methylphenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Methylphenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MDL Number: MFCD22546751
CAS Number: 1393568-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR