3-(4-Methylphenyl)-3-oxetanamine hydrochloride ≥95%

Supplier: Matrix Scientific

115919-1G 115919-5G 115919-10G
102898-802EA 1787.69 USD
102898-802 102898-804 102898-806
3-(4-Methylphenyl)-3-oxetanamine hydrochloride ≥95%
3-(4-Methylphenyl)-3-oxetanamine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClNO
MDL Number: MFCD19381737
CAS Number: 1322200-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR