Methyl(1-oxetan-3-ylpiperidin-4-yl)amine ≥95%

Supplier: Matrix Scientific

115918-250MG 115918-500MG 115918-1G
102898-796EA 963.08 USD
102898-796 102898-798 102898-800
Methyl(1-oxetan-3-ylpiperidin-4-yl)amine ≥95%
Methyl(1-oxetan-3-ylpiperidin-4-yl)amine

Hazard: Irritant

Formula: C₉H₁₈N₂O
MDL Number: MFCD21847920
CAS Number: 1416323-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR