3-Methylamino-3-hydroxymethyloxetane ≥95%

Supplier: Matrix Scientific

115914-250MG 115914-1G
102899-864EA 1593.85 USD
102899-864 102899-868
3-Methylamino-3-hydroxymethyloxetane ≥95%
3-Methylamino-3-hydroxymethyloxetane

Hazard: Irritant

Formula: C₅H₁₁NO₂
MDL Number: MFCD22628745
CAS Number: 1416323-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR