3-(3-Methoxyphenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115912-100MG 115912-250MG 115912-500MG
102898-790EA 1098.46 USD
102898-790 102898-990 102899-856
3-(3-Methoxyphenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Methoxyphenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₄
MDL Number: MFCD22546750
CAS Number: 1393583-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR