3-Methoxy-4-(oxetan-3-yloxy)benzonitrile ≥95%

Supplier: Matrix Scientific

115909-500MG 115909-1G 115909-5G
102898-742EA 1238.46 USD
102898-742 102898-774 102898-776
3-Methoxy-4-(oxetan-3-yloxy)benzonitrile ≥95%
3-Methoxy-4-(oxetan-3-yloxy)benzonitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD20233132
CAS Number: 1349717-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR