3-(2-Methoxy-4-nitrophenoxy)oxetane ≥95%

Supplier: Matrix Scientific

115907-250MG 115907-500MG
102898-736EA 963.08 USD
102898-736 102898-738
3-(2-Methoxy-4-nitrophenoxy)oxetane ≥95%
3-(2-Methoxy-4-nitrophenoxy)oxetane

Hazard: Irritant

Formula: C₁₀H₁₁NO₅
CAS Number: 1356113-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR