3-(Mercaptomethyl)-3-methyloxetane ≥95%

Supplier: Matrix Scientific

115906-500MG 115906-1G 115906-5G
102898-730EA 1044.62 USD
102898-730 102898-732 102898-734
3-(Mercaptomethyl)-3-methyloxetane ≥95%
3-(Mercaptomethyl)-3-methyloxetane

Hazard: Irritant

Formula: C₅H₁₀OS
MDL Number: MFCD01939890
CAS Number: 63991-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR