3-(3-Iodophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115904-100MG 115904-250MG 115904-500MG
102898-718EA 1718.46 USD
102898-718 102898-720 102898-722
3-(3-Iodophenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Iodophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉IO₃
MDL Number: MFCD22546749
CAS Number: 1393560-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR