3-(3-Iodophenyl)oxetan-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

115902-250MG 115902-500MG 115902-1G
102898-706EA 2267.69 USD
102898-706 102898-708 102898-710
3-(3-Iodophenyl)oxetan-3-amine hydrochloride ≥95%
3-(3-Iodophenyl)oxetan-3-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClINO
MDL Number: MFCD22628741
CAS Number: 1393560-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR