3-(3-Fluorophenyl)oxetane-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

115897-100MG 115897-250MG 115897-500MG
102898-676EA 1373.85 USD
102898-676 102898-678 102898-680
3-(3-Fluorophenyl)oxetane-3-carboxylic acid ≥95%
3-(3-Fluorophenyl)oxetane-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉FO₃
MDL Number: MFCD22546746
CAS Number: 1393553-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR