3-(3-Fluorophenyl)oxetan-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

115896-500MG 115896-1G 115896-5G
102898-670EA 1238.46 USD
102898-670 102898-672 102898-674
3-(3-Fluorophenyl)oxetan-3-amine hydrochloride ≥95%
3-(3-Fluorophenyl)oxetan-3-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₁ClFNO
MDL Number: MFCD19381727
CAS Number: 1332921-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR