[3-Fluoro-4-(oxetan-3-yloxy)phenyl]methanamine ≥95%

Supplier: Matrix Scientific

115894-250MG 115894-1G 115894-5G
102898-658EA 1238.46 USD
102898-658 102898-660 102898-662
[3-Fluoro-4-(oxetan-3-yloxy)phenyl]methanamine ≥95%
[3-Fluoro-4-(oxetan-3-yloxy)phenyl]methanamine

Hazard: Irritant

Formula: C₁₀H₁₂FNO₂
MDL Number: MFCD20233318
CAS Number: 1349709-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR