3-Fluoro-4-(oxetan-3-yloxy)benzoic acid ≥95%

Supplier: Matrix Scientific

115892-500MG 115892-1G 115892-5G
102898-646EA 1098.46 USD
102898-646 102898-648 102898-650
3-Fluoro-4-(oxetan-3-yloxy)benzoic acid ≥95%
3-Fluoro-4-(oxetan-3-yloxy)benzoic acid

Hazard: Irritant

Formula: C₁₀H₉FO₄
MDL Number: MFCD20233140
CAS Number: 1346498-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR