3-Fluoro-4-(oxetan-3-yloxy)aniline ≥95%

Supplier: Matrix Scientific

115891-10G 115891-5G
102898-644EA 4361.54 USD
102898-644 102899-848
3-Fluoro-4-(oxetan-3-yloxy)aniline ≥95%
3-Fluoro-4-(oxetan-3-yloxy)aniline

Hazard: Irritant

Formula: C₉H₁₀FNO₂
MDL Number: MFCD20233094
CAS Number: 1349716-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR