2-(3-Fluorooxetan-3-yl)ethanol ≥95%
Supplier: Matrix Scientific
115889-250MG
115889-500MG
102899-840EA
1200
USD
102899-840
102900-304
2-(3-Fluorooxetan-3-yl)ethanol ≥95%
2-(3-Fluorooxetan-3-yl)ethanol
Hazard: Irritant
Formula:
C₅H₉FO₂ |
MDL Number:
MFCD17215854 CAS Number: 1123786-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...