3-Ethynyl-3-methyloxetane 85-90%

Supplier: Matrix Scientific

115886-250MG 115886-500MG 115886-1G
102898-634EA 1415.38 USD
102898-634 102898-636 102898-638
3-Ethynyl-3-methyloxetane 85-90%
3-Ethynyl-3-methyloxetane

Hazard: Irritant

Formula: C₆H₈O
MDL Number: MFCD19441315
CAS Number: 1290541-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR