2,5-Dioxa-8-azaspiro[3.5]nonane oxalate ≥95%
Supplier: Matrix Scientific
115883-500MG
115883-1G
115883-250MG
102898-618EA
3309.23
USD
102898-618
102898-620
102898-772
2,5-Dioxa-8-azaspiro[3.5]nonane oxalate ≥95%
2,5-Dioxa-8-azaspiro[3.5]nonane oxalate
Hazard: Irritant
Formula:
C₈H₁₃NO₆ MW: 219.2 g/mol |
MDL Number:
MFCD22628735 CAS Number: 1184185-17-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...