4,4-Dimethyl-3-oxopentanal ≥97%

Supplier: Matrix Scientific

115882-1G 115882-5G 115882-25G
102898-766EA 70.77 USD
102898-766 102898-768 102898-770
4,4-Dimethyl-3-oxopentanal ≥97%
4,4-Dimethyl-3-oxopentanal

Hazard: Irritant

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00042806
CAS Number: 23459-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR